missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress NIM811

Product Code. 16472609
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16472609 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16472609 Supplier MedChemExpress Supplier No. HYP0025
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress NIM811 ((Melle-4)cyclosporin; SDZ NIM811) is an orally bioavailable mitochondrial permeability transition and cyclophilin dual inhibitor, which exhibits potent in vitro activity against hepatitis C virus (HCV) .

Specifications

Purity 98.02%
CAS 143205-42-9
Content And Storage Stored under nitrogen∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (stored under nitrogen)
For Use With (Application) COVID-19-anti-virus
Molecular Weight (g/mol) 1202.61 g/mol
Product Type Reference compound
Quantity 1 mg
Shipping Condition Room temperature
Solubility DMSO : 200 mg/mL (166.30 mM∣Need ultrasonic)
Synonym (Melle-4)cyclosporin∣ SDZ NIM811
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.