missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NIM1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113103
This item is not returnable.
View return policy
Description
NIM1 Polyclonal Antibody for Western Blot, IHC (P), ELISA
NIM1K serine/threonine kinase or NIM1 belongs to CAMK kinase family. Unlike to other AMPK-related kinases, NIM1 cannot be phosphorylated and activated by LKB1. Two somatic mutations (P333S and P411T) were found in lung neuroendocrine carcinoma and lung large cell carcinoma samples. NIM1 gene ontology (GO) annotations include cellular response to glucose starvation; intracellular signal transduction; negative regulation of TOR signaling; protein phosphorylation.
Specifications
| NIM1 | |
| Polyclonal | |
| Unconjugated | |
| NIM1K | |
| E130304F04Rik; Nim1; NIM1 serine/threonine protein kinase; NIM1 serine/threonine-protein kinase; NIM1K; Serine/threonine-protein kinase NIM1 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 167359 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 5 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q8IY84 | |
| NIM1K | |
| Recombinant Human Serine/threonine-protein kinase NIM1 protein (1-436AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction