missing translation for 'onlineSavingsMsg'
Learn More

Nilotinib, MedChemExpress

Product Code. 16430936
Encompass_Preferred
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
This item is not returnable. View return policy

Product Code. 16430936

Brand: Medchem Express HY10159

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Nilotinib is an orally available Bcr-Abl tyrosine kinase inhibitor with antineoplastic activity.

Nilotinib is an orally available Bcr-Abl tyrosine kinase inhibitor with antineoplastic activity.

Specifications

Chemical Name or Material Nilotinib
CAS 641571-10-0
Color Yellow
Purity Grade Notes Research
Molecular Formula C28H22F3N7O
Quantity 200 mg
Synonym AMN107
Solubility Information DMSO : < 1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble or slightly soluble)
SMILES O=C(NC1=CC(C(F)(F)F)=CC(N2C=NC(C)=C2)=C1)C3=CC=C(C)C(NC4=NC=CC(C5=CC=CN=C5)=N4)=C3
Molecular Weight (g/mol) 529.52
Formula Weight 529.52
Percent Purity 99.6%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.