missing translation for 'onlineSavingsMsg'
Learn More

Nile Red, TRC

Product Code. 30134657
Change view
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
10mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134657 25 mg 25mg
30008123 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134657 Supplier Toronto Research Chemicals Supplier No. TRCN46421825MG
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7385-67-3
Molecular Formula C20H18N2O2
Molecular Weight (g/mol) 318.37
Synonym 9-(Diethylamino)-5H-benzo[a]phenoxazin-5-one, A 84655, Nile Blue A Oxazone, Phenoxazone 9
IUPAC Name 9-(diethylamino)benzo[a]phenoxazin-5-one
SMILES BrCC1=CC(I)=CC=C1

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C20H18N2O2/c1-3-22(4-2)13-9-10-16-18(11-13)24-19-12-17(23)14-7-5-6-8-15(14)20(19)21-16/h5-12H,3-4H2,1-2H3
Formula Weight 318.1368
Chemical Name or Material Nile Red
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.