missing translation for 'onlineSavingsMsg'
Learn More

Nile red, MP Biomedicals™

Product Code. 11482502 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
250 mg
500 mg
Unit Size:
250mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11482502 250 mg 250mg
11492502 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11482502 Supplier MP Biomedicals Supplier No. 0215174483
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 7385-67-3
Molecular Formula C20H18N2O2
Molecular Weight (g/mol) 318.38
MDL Number MFCD00011639
InChI Key VOFUROIFQGPCGE-UHFFFAOYSA-N
Synonym Nile Blue A Oxazone, 9-Diethylamino-5H-benzo[a]phenoxazine-5-one
PubChem CID 65182
ChEBI CHEBI:52169
IUPAC Name 8-(diethylamino)-12H-10-oxa-5-azatetraphen-12-one
SMILES CCN(CC)C1=CC=C2N=C3C(OC2=C1)=CC(=O)C1=CC=CC=C31

Specifications

Formula Weight 318.4
Color Green
Quantity 250 mg
Physical Form Powder
Chemical Name or Material Nile red, Determination of lipids, staining whole tissue
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.