missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Nile Blue A, pure, certified

Product Code. p-3756579 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 10791822

Brand: Thermo Scientific Chemicals 415690100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3625-57-8 | C40H40N6O6S | 732.84 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3625-57-8
Molecular Formula C40H40N6O6S
Molecular Weight (g/mol) 732.84
MDL Number MFCD00064529
InChI Key QIRDPEPUXNCOLD-UHFFFAOYSA-N
Synonym Nile blue sulfate, C.I. 51180
PubChem CID 19256
IUPAC Name [9-(diethylamino)benzo[a]phenoxazin-5-ylidene]azanium;sulfate
SMILES CCN(CC)C1=CC2=C(C=C1)N=C3C4=CC=CC=C4C(=[NH2+])C=C3O2.CCN(CC)C1=CC2=C(C=C1)N=C3C4=CC=CC=C4C(=[NH2+])C=C3O2.[O-]S(=O)(=O)[O-]

Specifications

Absorption Ratio 1.01 to 1.16
Dye Content 70% min.
Beilstein 27, 404
Infrared Spectrum Authentic
Lambda Max 635 to 646nm
Formula Weight 732.87
Grade Pure
Packaging Glass bottle
Color Black to Green
Quantity 10 g
Physical Form Powder
Chemical Name or Material Nile Blue A, Pure
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.