missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ NIKi

Product Code. 30000013
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30000013 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30000013 Supplier Tocris Bioscience™ Supplier No. 7136/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent NIK inhibitor

NIKi is a potent inhibitor of NF-κB-inducing kinase (NIK) activity (IC50 = 20 nM).
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-[3-[2-Amino-5-(2-methoxyethoxy)-4-pyrimidinyl]-1H-indol-5-yl]-2-methyl-3-butyn-2-ol
CAS 1202763-91-4
Quantity 10 mg
Target NIK/MAP3K14
Molecular Formula C20H22N4O3
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.