missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NIK Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA141320
This item is not returnable.
View return policy
Description
Suggested positive control: antigen standard for MAP3K14 (transient overexpression lysate).
MAP3K14 (NIK) is structurally related to MAP3Ks. MAP3K14 phosphorylates and activates CHUK (IKKalpha) and IKBKB (IKK beta), which in turn phosphorylate IkB-alpha, releasing NF-KappaB leading to up-regulation of inflammatory response genes. This kinase binds to TRAF2 and stimulates NF-kappaB activity. It shares sequence similarity with several other MAPKK kinases. It participates in an NF-kappaB-inducing signalling cascade common to receptors of the tumour-necrosis/nerve-growth factor (TNF/NGF) family and to the interleukin-1 type-I receptor.
Specifications
| NIK | |
| Polyclonal | |
| Unconjugated | |
| MAP3K14 | |
| aly; alymphoplasia; FTDCR1B; HS; HsNIK; Map3k14; Mitogen-activated protein kinase kinase kinase 14; NF kappa B-inducing kinase; NF-kappa-beta-inducing kinase; Nfkb inducing kinase; NIK; protein kinase; serine/threonine-protein kinase NIK | |
| Rabbit | |
| Protein G | |
| RUO | |
| 9020 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q99558 | |
| MAP3K14 | |
| Synthetic peptide corresponding to amino acids 687-703 of human NIK. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction