missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NIK Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA5110933
This item is not returnable.
View return policy
Description
Immunogen sequence: EKNPSKASQS SRDTLSSGVH SWSSQAEARS SSWNMVLARG RPTDTPSYFN GVKVQIQSLN GEHLHIREFH RVKVGDIATG ISSQIPAAAF SLVTKDGQP
MAP3K14 encodes mitogen-activated protein kinase kinase kinase 14, which is a serine/threonine protein-kinase. This kinase binds to TRAF2 and stimulates NF-kappaB activity. It shares sequence similarity with several other MAPKK kinases. It participates in an NF-kappaB-inducing signalling cascade common to receptors of the tumour-necrosis/nerve-growth factor (TNF/NGF) family and to the interleukin-1 type-I receptor.
Specifications
| NIK | |
| Polyclonal | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q99558 | |
| MAP3K14 | |
| Recombinant Protein corresonding to Human NIK | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| Unconjugated | |
| MAP3K14 | |
| aly; BOS_19310; FTDCR1B; HS; HsNIK; MAP3K14; Mitogen-activated protein kinase kinase kinase 14; NF-kappa-beta-inducing kinase; NIK; Serine/threonine-protein kinase NIK | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9020 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction