missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NIK Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100732
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total NIK.
MAP3K14 (NIK) is structurally related to MAP3Ks. MAP3K14 phosphorylates and activates CHUK (IKKalpha) and IKBKB (IKK beta), which in turn phosphorylate IkB-alpha, releasing NF-KappaB leading to up-regulation of inflammatory response genes. This kinase binds to TRAF2 and stimulates NF-kappaB activity. It shares sequence similarity with several other MAPKK kinases. It participates in an NF-kappaB-inducing signalling cascade common to receptors of the tumour-necrosis/nerve-growth factor (TNF/NGF) family and to the interleukin-1 type-I receptor.
Specifications
| NIK | |
| Polyclonal | |
| Unconjugated | |
| MAP3K14 | |
| aly; alymphoplasia; FTDCR1B; HS; HsNIK; Map3k14; Mitogen-activated protein kinase kinase kinase 14; NF kappa B-inducing kinase; NF-kappa-beta-inducing kinase; Nfkb inducing kinase; NIK; protein kinase; serine/threonine-protein kinase NIK | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 53859, 9020 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q99558, Q9WUL6 | |
| MAP3K14 | |
| A synthesized peptide derived from human MAP3K14(Accession Q99558), corresponding to amino acid residues S787-A837. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction