missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ NIH 3T3 Whole Cell Lysate

Product Code. 18099108 Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
This item is not returnable. View return policy

Product Code. 18099108

Brand: Novus Biologicals™ NBP2039570.1MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

BALB/3T3 clone A31 (Mouse embryo) whole cell lysate 2mg/mL supplied in SDS sample buffer containing 5% Beta-mercaptoethanol. Growth media: DMEM and 10% FBS (fetal bovine serum). Lysates are guaranteed for 6 months from date of receipt.

The NIH 3T3 Whole Cell Lysate is derived from mouse. It has been validated for the following applications: Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Western Blot
Tissue NIH 3T3
Quantity 0.1 mg
Storage Buffer Supplied in SDS sample buffer containing 5% Beta-mercaptoethanol.
Test Specificity BALB/3T3 clone A31 (Mouse embryo)
Host Species Mouse
Concentration 2 mg/mL
Content And Storage Store at −80°C. Avoid freeze-thaw cycles.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.