missing translation for 'onlineSavingsMsg'
Learn More

NIH-12848, MedChemExpress

Product Code. 16469886
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16469886 50 mg 50mg
16463358 10 mg 10mg
16426087 5 mg 5mg
16424606 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16469886 Supplier Medchem Express Supplier No. HY101423
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress NIH-12848 is a putative phosphatidylinositol 5-phosphate 4-kinase γ (PI5P4Kγ) inhibitor with an IC50 of 1 μM.

NIH-12848 is a putative phosphatidylinositol 5-phosphate 4-kinase γ (PI5P4Kγ) inhibitor with an IC50 of 1 μM.

Specifications

Chemical Name or Material NIH-12848
CAS 959551-10-1
Color White
Purity Grade Notes Research
Molecular Formula C20H14F3N3S
Quantity 50 mg
Solubility Information DMSO : ≥ 100 mg/mL (259.46 mM)
SMILES FC(C1=CC=CC=C1C2=NC(NCC3=CC=CS3)=C4C=CC=CC4=N2)(F)F
Molecular Weight (g/mol) 385.41
Formula Weight 385.41
Percent Purity 98.06%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.