missing translation for 'onlineSavingsMsg'
Learn More

Nigrosin, soluble in spirit, Thermo Scientific™

Product Code. 11459397
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11459397 25 g 25g
11469397 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11459397 Supplier Thermo Scientific Alfa Aesar Supplier No. B21424.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11099-03-9
Molecular Formula C8H19ClN2O
Molecular Weight (g/mol) 194.703
MDL Number MFCD00149005
InChI Key GBPWTMZRCAESKB-UHFFFAOYSA-N
Synonym 2-amino-n,n-dipropylacetamide hydrochloride, solvent black 5, nigrosine spirit soluble solvent black 5 and its salts, when used as a substance in hair dye products
PubChem CID 19828004
IUPAC Name 2-amino-N,N-dipropylacetamide;hydrochloride
SMILES CCCN(CCC)C(=O)CN.Cl

Specifications

Melting Point >300°C
Quantity 25 g
Solubility Information Soluble in spirit
Chemical Name or Material Nigrosin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.