missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nigericin sodium salt, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Description
Nigericin sodium salt is an antibiotic from Streptomyces hygroscopicus that works by acting as an H+, K+, and Pb2+ ionophore, a NLRP3 activator.
Specifications
Specifications
| Chemical Name or Material | Nigericin sodium salt |
| Physical Form | Powder |
| CAS | 28643-80-3 |
| Color | White |
| Quantity | 25 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C40H67NaO11 |
| For Use With (Application) | Cancer-programmed cell death |
| Solubility Information | Ethanol : ≥ 50 mg/mL (66.94 mM) |
| SMILES | C[C@H]1[C@](O[C@@H]2C[C@](CC[C@@H]3C)([H])O[C@@]3([H])[C@@H](C)C(O[Na])=O)([C@@H]([C@H](OC)C2)C)O[C@](C)([C@@]4([H])O[C@@](C)([C@@]5([H])O[C@@]([C@]([C@@H](C[C@@H]6C)C)([H])O[C@]6(O)CO)([H])C[C@H]5C)CC4)C1 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction