missing translation for 'onlineSavingsMsg'
Learn More

Nigakinone, MedChemExpress

Product Code. 16451678
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16451678 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16451678 Supplier Medchem Express Supplier No. HYN2128

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Nigakinone is one of the most abundant alkaloids responsible for the major pharmacological activities of Kumu.

Nigakinone is one of the most abundant alkaloids responsible for the major pharmacological activities of Kumu.

Specifications

Chemical Name or Material Nigakinone
CAS 18110-86-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C15H10N2O3
Quantity 5 mg
Solubility Information DMSO : 8.33 mg/mL (31.29 mM; Need ultrasonic)
SMILES O=C1N2C3=C(N=CC=C3C4=C2C=CC=C4)C(OC)=C1O
Molecular Weight (g/mol) 266.25
Formula Weight 266.25
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.