missing translation for 'onlineSavingsMsg'
Learn More

Nifursol, MedChemExpress

Product Code. 16468796
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16468796 100 mg 100mg
16432286 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16468796 Supplier Medchem Express Supplier No. HYB1703
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Nifursol is a potent and orally active veterinary antibiotic for the prevention of histomoniasis. Nifursol rapidly metabolizes to form the metabolic marker 3,5-dinitrosalicyclic acid hydrazide (DNSAH) which can persist for a long time. Nifursol is widely used for the research of Escherichia Gastroenteropathy in poultry, fowl and aquatic animal.

Nifursol is a potent and orally active veterinary antibiotic for the prevention of histomoniasis. Nifursol rapidly metabolizes to form the metabolic marker 3,5-dinitrosalicyclic acid hydrazide (DNSAH) which can persist for a long time. Nifursol is widely used for the research of Escherichia Gastroenteropathy in poultry, fowl and aquatic animal.

Specifications

Chemical Name or Material Nifursol
Physical Form Solid
CAS 16915-70-1
Color Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C12H7N5O9
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 50 mg/mL (136.91 mM; Need ultrasonic)
SMILES O=C(N/N=C/C1=CC=C([N+]([O-])=O)O1)C2=CC([N+]([O-])=O)=CC([N+]([O-])=O)=C2O
Molecular Weight (g/mol) 365.21
Formula Weight 365.21
Percent Purity 97.8%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.