missing translation for 'onlineSavingsMsg'
Learn More

Nifuroxazide, MedChemExpress

Product Code. 16461456
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 mg
500 mg
Unit Size:
200mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16461456 200 mg 200mg
16452276 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16461456 Supplier Medchem Express Supplier No. HYB1436
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Nifuroxazide is an effective inhibitor of STAT3, also exerts potent anti-tumor and anti-metastasis activity.

Nifuroxazide is an effective inhibitor of STAT3, also exerts potent anti-tumor and anti-metastasis activity.

Specifications

Chemical Name or Material Nifuroxazide
CAS 965-52-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C12H9N3O5
Quantity 200 mg
Solubility Information DMSO : ≥ 155 mg/mL (563.19 mM)
SMILES O=C(N/N=C/C1=CC=C([N+]([O-])=O)O1)C2=CC=C(O)C=C2
Molecular Weight (g/mol) 275.22
Formula Weight 275.22
Percent Purity 98.51%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.