missing translation for 'onlineSavingsMsg'
Learn More

Nifuratel, MedChemExpress

Product Code. p-200101780
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16498006 100 mg 100mg
16484167 50 mg 50mg
16408288 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16498006 Supplier Medchem Express Supplier No. HYA0059

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Nifuratel(NF 113, SAP 113) is a broad antibacterial spectrum agent, which is used as an antibacterial, antifungal, and antiprotozoal (Trichomonas).

Nifuratel(NF 113, SAP 113) is a broad antibacterial spectrum agent, which is used as an antibacterial, antifungal, and antiprotozoal (Trichomonas).

Specifications

Chemical Name or Material Nifuratel
Physical Form Solid
CAS 4936-47-4
Color Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C10H11N3O5S
For Use With (Application) COVID-19-immunoregulation
Synonym NF 113 SAP 113 Methylmercadone
Solubility Information DMSO : 50 mg/mL (175.27 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C1OC(CSC)CN1/N=C/C2=CC=C([N+]([O-])=O)O2
Molecular Weight (g/mol) 285.28
Formula Weight 285.28
Percent Purity 98.87%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.