missing translation for 'onlineSavingsMsg'
Learn More

Niflumic acid, MedChemExpress

Product Code. 16408346
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16408346 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16408346 Supplier Medchem Express Supplier No. HYB0493
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Niflumic acid, a Ca2+-activated Cl- channel blocker, is an analgesic and anti-inflammatory agent used in the treatment of rheumatoid arthritis.

Niflumic acid, a Ca2+-activated Cl- channel blocker, is an analgesic and anti-inflammatory agent used in the treatment of rheumatoid arthritis.

Specifications

Chemical Name or Material Niflumic acid
CAS 4394-00-7
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C13H9F3N2O2
Quantity 100 mg
Solubility Information DMSO : ≥ 100 mg/mL (354.33 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(C1=CC=CN=C1NC2=CC=CC(C(F)(F)F)=C2)O
Molecular Weight (g/mol) 282.22
Formula Weight 282.22
Percent Purity 99.84%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.