missing translation for 'onlineSavingsMsg'
Learn More

Niflumic acid, 99+%

Product Code. 15493259
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15493259 10 g 10g
15483259 5 g 5g
15403269 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15493259 Supplier Thermo Scientific Alfa Aesar Supplier No. J60489.09
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4394-00-7 | C13H9F3N2O2 | 282.22 g/mol

It is used as a selective Cox-2 inhibitor and GPR35 activator. Also activates human TRPA1 inducibly expressed in HEK293 cells; displays agonist activity at GPR35.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a selective Cox-2 inhibitor and GPR35 activator. Also activates human TRPA1 inducibly expressed in HEK293 cells; displays agonist activity at GPR35.

Solubility
Soluble in ethanol (∼50 mg/ml), acetone (50 mg/ml, Clear to Slightly Hazy, Yellow), methanol (∼50 mg/ml), DMSO (56 mg/ml at 25°C), and acetonitrile (∼50 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Light sensitive. Hygroscopic. Keep away fom strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4394-00-7
Molecular Formula C13H9F3N2O2
Molecular Weight (g/mol) 282.22
MDL Number MFCD00010569
InChI Key JZFPYUNJRRFVQU-UHFFFAOYSA-N
Synonym niflumic acid, nifluril, landruma, forenol, actol, acido niflumico, nifluminic acid, acide niflumique, acidum niflumicum, acide niflumique inn-french
PubChem CID 4488
ChEBI CHEBI:34888
IUPAC Name 2-{[3-(trifluoromethyl)phenyl]amino}pyridine-3-carboxylic acid
SMILES OC(=O)C1=CC=CN=C1NC1=CC=CC(=C1)C(F)(F)F

Specifications

Melting Point 203°C to 204°C
Color Yellow
Quantity 10 g
UN Number UN2811
Sensitivity Light sensitive
Merck Index 14,6531
Solubility Information Soluble in ethanol (∽50mg/ml),acetone (50mg/ml,Clear to Slightly Hazy,Yellow),methanol (∽50mg/ml),DMSO (56mg/ml at 25°C),and acetonitrile (∽50mg/ml).
Formula Weight 282.22
Percent Purity ≥99%
Physical Form Crystalline Powder
Chemical Name or Material Niflumic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.