missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NIFK Antibody (CL2237) [DyLight 350], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-36750UV
This item is not returnable.
View return policy
Description
NIFK Monoclonal antibody specifically detects NIFK in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| NIFK | |
| Monoclonal | |
| DyLight 350 | |
| hNIFK, MKI67 (FHA domain) interacting nucleolar phosphoprotein, MKI67 FHA domain-interacting nucleolar phosphoprotein, NIFKNOPP34, Nopp34, Nucleolar phosphoprotein Nopp34, Nucleolar protein interacting with the FHA domain of pKI-67 | |
| This antibody was developed using a recombinant protein derived from Q9BYG3, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 84365 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL2237 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction