missing translation for 'onlineSavingsMsg'
Learn More

Nifedipine, MedChemExpress

Product Code. 16462119
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16462119 1 g 1g
16450858 5 g 5g
16408448 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16462119 Supplier Medchem Express Supplier No. HYB0284
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Nifedipine (BAY-a-1040) is a potent calcium channel blocker and drug of choice for cardiac insufficiencies.

Nifedipine (BAY-a-1040) is a potent calcium channel blocker and drug of choice for cardiac insufficiencies.

Specifications

Chemical Name or Material Nifedipine
CAS 21829-25-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C17H18N2O6
Quantity 1 g
Synonym BAY-a-1040
Solubility Information DMSO : 100 mg/mL (288.74 mM; Need ultrasonic) ∣H2O : 1 mg/mL (2.89 mM; ultrasonic and warming and heat to 80°C)
SMILES O=C(C1=C(C)NC(C)=C(C(OC)=O)C1C2=CC=CC=C2[N+]([O-])=O)OC
Molecular Weight (g/mol) 346.33
Formula Weight 346.33
Percent Purity 99.35%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.