missing translation for 'onlineSavingsMsg'
Learn More

Nifedipine, LoGiCal

Product Code. 30459030
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30459030 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30459030 Supplier LoGiCal Supplier No. LGCFOR0003.00
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nifedipine
CAS 21829-25-4
Analyte or Component Names Nifedipine
Shipping Condition Room Temperature
Molecular Formula C17 H18 N2 O6
Certifications/Compliance ISO 9001
Synonym 3,5-Pyridinedicarboxylic acid, 1,4-dihydro-2,6-dimethyl-4-(2-nitrophenyl)-, 3,5-dimethyl ester, 3,5-Pyridinedicarboxylic acid, 1,4-dihydro-2,6-dimethyl-4-(2-nitrophenyl)-, dimethyl ester, 3,5-Pyridinedicarboxylic acid, 1,4-dihydro-2,6-dimethyl-4-(o-nitrophenyl)-, dimethyl ester, 2,6-Dimethyl-3,5-dicarbomethoxy-4-(2-nitrophenyl)-1,4-dihydropyridine, 2,6-Dimethyl-4-(2-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylic acid dimethyl ester, 4-(2-Nitrophenyl)-2,6-dimethyl-3,5-dicarbomethoxy-1,4-dihydropyridine, Adalat, Adalat 10, Adalat 20, Adalat 5, Adalat CC, Adalat CR, Adalat Crono, Adalat FT, Adalat GITS, Adalat GITS 30, Adalat LA, Adalat LP, Adalat Oros, Adalat PA, Adalat retard, Adalate, Adapine, Adapress, Alat, Aldipin, Alfadat, Alonix, Alonix S, Alpha-Nifedipine Retard, Angipec, Anifed, Anpine, Apo-Nifed, Aprical, BAY 1040, BAY-a 1040, Bonacid, Calcibloc, Calcigard, Calcilat, Camont, Cardifen, Cardilat, Cardilate, Cardionorm, Chronadalate, Chronadalate LP, Citilat, Coracten, Coral, Cordafen, Cordaflex, Cordalat, Cordicant, Cordilan, Cordipin, Corinfar, Corotrend, Corynphar, Depin, Dignokonstant, Dilafed, Dilcor, Dimethyl 1,4-dihydro-2,6-dimethyl-4-(2-nitrophenyl)-3,5-pyridinedicarboxylate, Dimethyl 4-(o-Nitrophenyl)-2,6-dimethyl-1,4-dihydro-3,5-pyridinedicarboxylate, Dipinkor, Duranifin, Ecodipi, Ecodipin, Ecodipin E, Fedcor, Fedcor Retard, Fedipin, Fenamon, Fenamon SR, Fenihidine, Glopir, Hadipine, Hexadilat, Introcar, Kordafen, Macorel, Megalat, Myogard, NIfedilat, Nedipin, Nicardia, Nifangin, Nifar, Nifdemin, Nifebene, Nifecard, Nifecor, Nifedate, Nifedepat, Nifedicor, Nifedin, Nifedine, Nifedipine, Nifedipine retard, Nifedipres, Nifedirex LP, Nifelan, Nifelat, Nifelat Q, Nifelate, Nifensar XL, Nificard, Nifidine, Nifipen, Niphedipine, Normadil, Novo Nifedin, Nyefax, Nyefax Retard, Orix, Osmo-Adalat, Oxcord, Pidilat, Procardia, Procardia XL, Sepamit, Sepamit R, Slofedipine XL, TN 873R, Tensopin, Tibricol, UV-ENP 349PINA, Vasdalat, Vasdalat Retard, Vasofed, Zenusin
InChI Formula InChI=1S/C17H18N2O6/c1-9-13(16(20)24-3)15(14(10(2)18-9)17(21)25-4)11-7-5-6-8-12(11)19(22)23/h5-8,15,18H,1-4H3
Solution Type Neat
SMILES COC(=O)C1=C(C)NC(=C(C1c2ccccc2[N+](=O)[O-])C(=O)OC)C
IUPAC Name dimethyl 2,6-dimethyl-4-(2-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
Industry Type Clinical Testing
Molecular Weight (g/mol) 346.33
Quantity 10 mg
Formula Weight 346.1165 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.