missing translation for 'onlineSavingsMsg'
Learn More

Nifedipine, 98%, Thermo Scientific™

L-type calcium channel blocker; induces apoptosis in human glioblastoma cells

£60.65 - £455.00

Chemical Identifiers

CAS 21829-25-4
Molecular Formula C17H18N2O6
Molecular Weight (g/mol) 346.34
MDL Number MFCD00057326
InChI Key HYIMSNHJOBLJNT-UHFFFAOYSA-N
Synonym nifedipine, procardia, adalat, procardia xl, adalat cc, cordipin, corinfar, fenihidine, citilat, oxcord
PubChem CID 4485
ChEBI CHEBI:7565
IUPAC Name 3,5-dimethyl 2,6-dimethyl-4-(2-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
SMILES COC(=O)C1=C(C)NC(C)=C(C1C1=CC=CC=C1[N+]([O-])=O)C(=O)OC
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15495109
SDS View Documents
Thermo Scientific Alfa Aesar
J62811.06
5g
ONLINE EXCLUSIVE PRICE
£67.50
£60.65 / 5g
Save £6.85
10% Off
View alternative products
Save Up to 
»
Estimated Shipment: 28-09-2026
Add to basket
 
 Restricted Item
15405119
SDS View Documents
Thermo Scientific Alfa Aesar
J62811.14
£290.00
25g
View alternative products
Save Up to 
»
Estimated Shipment: 28-09-2026
Add to basket
 
 Restricted Item
15415119
SDS View Documents
Thermo Scientific Alfa Aesar
J62811.22
100g
£455.00
100g
View alternative products
Save Up to 
»
Estimated Shipment: 28-09-2026
Add to basket
 
 Restricted Item
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
L-type calcium channel blocker; induces apoptosis in human glioblastoma cells.Nifedipine is a dihydropyridine calcium channel blocker used as L-type calcium channel blocker, antianginal and an antihypertensive . It induces apoptosis in human glioblastoma cells. It is used as a coronary vasodilator in the treatment of cardiac and circulatory disorders. Further, it is used in high-altitude medicine to treat high altitude pulmonary edema, hypertension and chronic stable angina. In addition, it is frequently used as a tocolytic agent.

Solubility
Soluble in acetone, methylene chloride, chloroform, ethyl acetate, dimethyl sulfoxide and methanol. Sparingly soluble in ethanol. Insoluble in water.

Notes
Light sensitive. Store in a cool place. Incompatible with strong acids, strong oxidizing agents, caustics, aliphatic amines, amides, diethylamine, potassium hydride and potassium hydroxide.
Specifications

Chemical Identifiers

21829-25-4
346.34
HYIMSNHJOBLJNT-UHFFFAOYSA-N
4485
3,5-dimethyl 2,6-dimethyl-4-(2-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
C17H18N2O6
MFCD00057326
nifedipine, procardia, adalat, procardia xl, adalat cc, cordipin, corinfar, fenihidine, citilat, oxcord
CHEBI:7565
COC(=O)C1=C(C)NC(C)=C(C1C1=CC=CC=C1[N+]([O-])=O)C(=O)OC

Specifications

21829-25-4
MFCD00057326
nifedipine, procardia, adalat, procardia xl, adalat cc, cordipin, corinfar, fenihidine, citilat, oxcord
HYIMSNHJOBLJNT-UHFFFAOYSA-N
3,5-dimethyl 2,6-dimethyl-4-(2-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
4485
346.33
Nifedipine
C17H18N2O6
14,6528
Soluble in acetone,methylene chloride,chloroform,ethyl acetate,dimethyl sulfoxide and methanol. Sparingly soluble in ethanol. Insoluble in water.
COC(=O)C1=C(C)NC(C)=C(C1C1=CC=CC=C1[N+]([O-])=O)C(=O)OC
346.34
CHEBI:7565
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

missing translation for 'einecsNumber' : 244-598-3

missing translation for 'rtecsNumber' : US7975000

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

Product Identifier
  • Nifedipine

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H302-Harmful if swallowed.

Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

Supplemental information
  • MIXTURE LIST-Contain: Nifedipine

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.