missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NIF3L1 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33552-100ul
This item is not returnable.
View return policy
Description
NIF3L1 Monoclonal antibody specifically detects NIF3L1 in Human,Rat samples. It is validated for ELISA,Western Blot
Specifications
| NIF3L1 | |
| Monoclonal | |
| Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL | |
| ALS2CR1, candidate 1, MDS015, NIF3 NGG1 interacting factor 3-like 1 (S. pombe), NIF3-like protein 1, S.pombe homolog)-like 1 | |
| A synthetic peptide corresponding to a sequence within amino acids 250-350 of human NIF3L1 (Q9GZT8).,, Sequence:, MGRLCTLDESVSLATMIDRIKRHLKLSHIRLALGVGRTLESQVKVVALCAGSGSSVLQGVEADLYLTGEMSHHDTLDAASQGINVILCEHSNTERGFLSDL | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 60491 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction