missing translation for 'onlineSavingsMsg'
Learn More

NIF1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NB100-2579

Product Code. 18015685

  • £452.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

NIF1 Polyclonal specifically detects NIF1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NIF1
Polyclonal
Unconjugated
TBS and 0.1% BSA with 0.09% Sodium Azide
DKFZp586G1122, Hzf, HZFzinc finger protein 385, RZFHematopoietic zinc finger protein, ZFP385, zinc finger protein 385A, ZNF385Retinal zinc finger protein
Rabbit
Affinity Purified
RUO
63925
Store at 4C. Do not freeze.
Western Blot, Immunohistochemistry, Immunoprecipitation
0.2 mg/mL
Western Blot 1:2000-1:10000, Immunohistochemistry, Immunoprecipitation 2-5 ug/mg lysate
NP_071378.1
ZNF335
The immunogen recognized by this antibody maps to a region between residue 1300 and the C-terminus (residue 1342) of human NRC-Interacting Factor 1 using the numbering given in entry NP_071378.1 (GeneID 63925).
0.1 mL
Primary
Human, Mouse
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only