missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Niemann-Pick C1 Antibody (1318A) [Janelia Fluor™ 669], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-76798JF669
This item is not returnable.
View return policy
Description
Niemann-Pick C1 Monoclonal antibody specifically detects Niemann-Pick C1 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Gene Knock-Out,Immunocytochemistry/ Immunofluorescence
Specifications
| Niemann-Pick C1 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| FLJ98532, Niemann-Pick C1 protein, Niemann-Pick disease, type C1, NPC, NPC1 | |
| A synthetic peptide made to the C-terminal region of human Niemann-Pick C. [UniProt# O15118] | |
| 0.1 mL | |
| Cancer, Cholesterol Metabolism, Lipid and Metabolism, Membrane Trafficking and Chaperones, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Neurotransmission, Signal Transduction | |
| 4864 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Gene Knock-Out, Immunocytochemistry/Immunofluorescence | |
| 1318A | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction