missing translation for 'onlineSavingsMsg'
Learn More

Niemann-Pick C1 Antibody (1318A), DyLight 680, Novus Biologicals™

Product Code. 18430008 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18430008 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18430008 Supplier Novus Biologicals Supplier No. NBP276798FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Niemann-Pick C1 Monoclonal antibody specifically detects Niemann-Pick C1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Gene Knock-Out
TRUSTED_SUSTAINABILITY

Specifications

Antigen Niemann-Pick C1
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Gene Knock-Out
Classification Monoclonal
Clone 1318A
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias FLJ98532, Niemann-Pick C1 protein, Niemann-Pick disease, type C1, NPC, NPC1
Host Species Rabbit
Immunogen A synthetic peptide made to the C-terminal region of human Niemann-Pick C. [UniProt# O15118]
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cholesterol Metabolism, Lipid and Metabolism, Membrane Trafficking and Chaperones, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Neurotransmission, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4864
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.