missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Nidogen Recombinant Rabbit Monoclonal Antibody (K01_2M26)

Product Code. 30830286
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30830286 50 μL
30825608 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30830286 Supplier Invitrogen™ Supplier No. MA565514

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: VEKTRCQHER EHILGAAGAT DPQRPIPPGL FVPECDAHGH YAPTQCHGST GYCWCVDRDG REVEGTRTRP GMTPPCLSTV APPIHQGPAV PTAVIPLPPG THLLFAQTGK IERLPLEGNT MRKTEAKAFL HVPAKVIIGL AFDCVDKMVY WTDITEPSIG RASLHGGEPT TIIRQDLGSP EGIAVDHLGR NIFWTDSNLD RIEVAKLDGT QRRVLFETDL VNPRGIVTDS VRGNLYWTDW NRDNPKIETS YMDGTNRRIL VQDDLGLPNG LTFDAFSSQL CWVDAGTNRA ECLNPSQPSR RKALEGLQYP FAVTSYGKNL YFTDWKMNSV VALDLAISKE TDAFQPHKQT RLYGITTALS QCPQGHNYCS VNNGGCTHLC LATPGSRTCR CPDNTLGVDC IE Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 50%, Pig - 91%, Cynomolgus monkey - 98%.

This gene encodes a member of the nidogen family of basement membrane glycoproteins. The protein interacts with several other components of basement membranes, and may play a role in cell interactions with the extracellular matrix.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nidogen
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone K01_2M26
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 40% glycerol, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene NID1
Gene Accession No. P14543
Gene Alias A630025O17; enactin; Ent; Entactin; entactin 1; entactin-1; Nid; NID 1; NID1; NID-1; Nidogen (enactin); nidogen (entactin); nidogen 1; Nidogen precursor (Entactin); nidogen-1; Nidogen-1;
Gene Symbols NID1
Host Species Rabbit
Immunogen Recombinant protein of human Entactin/NID (aa 844-1245).
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4811
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.