missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Nidogen Recombinant Rabbit Monoclonal Antibody (24GB4995)

Rabbit Recombinant Monoclonal Antibody

£126.00 - £434.00

Specifications

Antigen Nidogen
Clone 24GB4995
Concentration 0.7 mg/mL
Content And Storage -20°C
Applications Flow Cytometry, Western Blot
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30276068
View Documents
Invitrogen™
MA557119
100 μL
£434.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30274861
View Documents
Invitrogen™
MA557120
20 μL
£126.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Nidogen Recombinant Monoclonal Antibody for Western Blot, Flow

This gene encodes a member of the nidogen family of basement membrane glycoproteins. The protein interacts with several other components of basement membranes, and may play a role in cell interactions with the extracellular matrix.
Specifications

Specifications

Nidogen
0.7 mg/mL
Flow Cytometry, Western Blot
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P08460, P10493
18073, 25494
Recombinant protein of human Entactin/NID (aa 844-1245).
Antibody
24GB4995
-20°C
Recombinant Monoclonal
Liquid
IgG
Mouse, Rat
NID1
A630025O17; enactin; Ent; Entactin; entactin 1; entactin-1; Nid; NID 1; NID1; NID-1; Nidogen (enactin); nidogen (entactin); nidogen 1; Nidogen precursor (Entactin); nidogen-1; Nidogen-1;
NID1
Primary
Affinity chromatography
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.