missing translation for 'onlineSavingsMsg'
Learn More

Nidogen G3 Domain Monoclonal Antibody (JF2)
GREENER_CHOICE

Nidogen G3 Domain Monoclonal Antibody (JF2)

Brand:  Invitrogen MA517020

 View more versions of this product

Product Code. 15388520

  • £580.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Nidogen G3 Domain
Monoclonal
1 mg/mL
0.5M acetic acid, 2M tris with 0.1% sodium azide
P10493, P14543
NID1
Recombinant fragment corresponding to the G3 region of mouse nidogen produced in Fisher rat plus YB2/0 cells.
100 μL
Primary
Human, Mouse
Antibody
IgG2a
ELISA, Inhibition Assays, Western Blot
JF2
Unconjugated
NID1
A630025O17; enactin; Ent; Ent1; Entactin; entactin 1; entactin-1; NID; NID 1; Nid1; NID-1; Nidogen (enactin); nidogen 1; Nidogen precursor (Entactin); nidogen-1
Rat
Protein G
RUO
18073, 4811
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.