Learn More
Invitrogen™ Nidogen G2 Domain Monoclonal Antibody (JF4)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA517017
Description
MA5-17017 recognizes native and denatured mouse and human nidogen, and blocks binding of nidogen to laminin. MA5-17017 has been successfully used in ELISA and Western blot applications. The MA5-17017 immunogen is a recombinant fragment corresponding to the G2 region of mouse nidogen produced in Fisher rat plus YB2/0 cells.
Basement membrane components play an important role in cell adhesion, locomotion, differentiation and embryonic compartmentalization. Although the precise composition and structure of basal laminae varies from tissue to tissue, as well as within the same tissue at different developmental stages, most mature basal laminae contain type IV collagen, laminins, heparan sulphate proteoglycans and entactin (or nidogen). Entactin is a sulfated glycoprotein that is widely distributed in basement membranes. It is tightly associated with laminin and also binds to collagen IV. Entactin probably has a role in cell-extracellular matrix interactions.
Specifications
| Nidogen G2 Domain | |
| Monoclonal | |
| 1 mg/mL | |
| 0.5M acetic acid, 2M tris with 0.1% sodium azide | |
| P10493, P14543 | |
| NID1 | |
| Recombinant fragment corresponding to the G2 region of mouse nidogen produced in Fisher rat plus YB2/0 cells. | |
| 50 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG2b |
| ELISA, Inhibition Assays, Western Blot | |
| JF4 | |
| Unconjugated | |
| NID1 | |
| A630025O17; enactin; Ent; Ent 2; entactin; entactin 1; Entactin 2; entactin-1; NID; NID 2; NID1; NID-1; Nid2; Nidogen (enactin); nidogen 1; Nidogen 2; Nidogen precursor (Entactin); nidogen-1 | |
| Rat | |
| Protein G | |
| RUO | |
| 18073, 4811 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.