missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nidogen 2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13530-1-AP-150UL
This item is not returnable.
View return policy
Description
Basement membranes, which are composed of type IV collagens, laminins, perlecan, and nidogen, are thin pericellular protein matrices that control a large number of cellular activities, including adhesion, migration, differentiation, gene expression, and apoptosis. Basement membranes, which are composed of type IV collagens, perlecan, and nidogen, are thin pericellular protein matrices that control a large number of cellular activities, including adhesion, migration, differentiation, gene expression, and apoptosis.Specifications
| Nidogen 2 | |
| Polyclonal | |
| Unconjugated | |
| NID2 | |
| NID 2, NID2, Nidogen 2, nidogen 2 (osteonidogen), Osteonidogen | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 18074, 22795, 302248 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| B5DFC9, O88322, Q14112 | |
| Nid2 | |
| Nidogen 2 Fusion Protein Ag4424 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction