missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nidogen 2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA562615
This item is not returnable.
View return policy
Description
Immunogen sequence: HTKEGTSLGE VGGPDLKGQV EPWDERETRS PAPPEVDRDS LAPSWETPPP YPENGSIQPY PDGGPVPSEM DVPPAHPEEE IVLRSYPASG HTT Highest antigen sequence identity to the following orthologs: Mouse - 43%, Rat - 44%.
Basement membranes, which are composed of type IV collagens, laminins, perlecan, and nidogen, are thin pericellular protein matrices that control a large number of cellular activities, including adhesion, migration, differentiation, gene expression, and apoptosis.Basement membranes, which are composed of type IV collagens (see MIM 120130), laminins (see LAMC1; MIM 150290), perlecan (HSPG2; MIM 142461), and nidogen (see NID1; MIM 131390), are thin pericellular protein matrices that control a large number of cellular activities, including adhesion, migration, differentiation, gene expression, and apoptosis.[supplied by OMIM].
Specifications
| Nidogen 2 | |
| Polyclonal | |
| Unconjugated | |
| NID2 | |
| Nidogen 2 (osteonidogen), NID-2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 22795 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry | |
| 0.5 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q14112 | |
| NID2 | |
| Recombit protein corresponding to Human Nidogen 2 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction