missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Nidogen-1/NID1 Human ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ EHNID1X10
Additional Details : Weight : 2.50000kg
Description
This gene encodes a member of the nidogen family of basement membrane glycoproteins. The protein interacts with several other components of basement membranes, and may play a role in cell interactions with the extracellular matrix.Specifications
P14543 | |
100 pg/mL | |
ELISA | |
Colorimetric Microplate Reader | |
NID1 | |
<10% | |
HRP | |
RUO | |
2°C to 8°C | |
1 hr 20 min |
0.123 to 30 ng/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
4811 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
10 x 96 Tests | |
Plasma, 2 μL|Serum, 2 μL|Supernatant, 100 μL | |
Nidogen | |
4 hr 45 min |