missing translation for 'onlineSavingsMsg'
Learn More

Nidogen-1/Entactin Antibody (ELM1), DyLight 650, Novus Biologicals™

Product Code. 18400228 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18400228 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18400228 Supplier Novus Biologicals Supplier No. NBP197701C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

Nidogen-1/Entactin Monoclonal antibody specifically detects Nidogen-1/Entactin in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Frozen)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nidogen-1/Entactin
Applications Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone ELM1
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias enactin, Entactin, NID-1, NIDentactin, nidogen (enactin), nidogen 1, nidogen-1
Host Species Rat
Immunogen Derived by fusion of X63 Ag 8.653 mouse myeloma cells with spleen cells from a Fisher rat immunized with a partially purified preparation of laminin from the EHS mouse tumor.
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4811
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.