missing translation for 'onlineSavingsMsg'
Learn More

Nidogen-1/Entactin Antibody (ELM1) - Azide and BSA Free, Novus Biologicals™

Product Code. 18326342 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18326342 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18326342 Supplier Novus Biologicals Supplier No. NBP308237

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

Nidogen-1/Entactin Monoclonal specifically detects Nidogen-1/Entactin in Mouse, Guinea Pig samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nidogen-1/Entactin
Applications ELISA
Classification Monoclonal
Clone ELM1
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution ELISA : 2-4 ug/mL
Formulation 10mM PBS
Gene Alias enactin, Entactin, NID-1, NIDentactin, nidogen (enactin), nidogen 1, nidogen-1
Host Species Rat
Immunogen Murine EHS laminin preparation
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4811
Target Species Mouse, Guinea Pig
Content And Storage Store at -20 to -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.