missing translation for 'onlineSavingsMsg'
Learn More

Nicotinuric Acid, Mikromol™

Product Code. 30052156
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30052156 100 mg 100mg
30051303 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30052156 Supplier Mikromol Supplier No. MM0553.13

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Nicotinuric Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nicotinuric Acid
CAS 583-08-4
Analyte or Component Names Nicotinuric Acid
Shipping Condition Room Temperature
Molecular Formula C8 H8 N2 O3
Certifications/Compliance ISO 9001
Synonym Glycine, N-(3-pyridinylcarbonyl)-, Glycine, N-nicotinoyl- (6CI,7CI,8CI), (Nicotinoylamino)acetic acid, 2-(Nicotinylamino)acetic acid, NSC 71273, Nicotinoylglycine, Nicotinuric acid, Nicotinylglycine
InChI Formula InChI=1S/C8H8N2O3/c11-7(12)5-10-8(13)6-2-1-3-9-4-6/h1-4H,5H2,(H,10,13)(H,11,12)
SMILES OC(=O)CNC(=O)c1cccnc1
IUPAC Name 2-(pyridine-3-carbonylamino)acetic acid
Molecular Weight (g/mol) 180.16
Quantity 100 mg
Formula Weight 180.0535 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.