missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nicotinuric acid, MedChemExpress
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Nicotinuric acid is an acyl glycine. Nicotinuric acid is a metabolite of nicotinic acid.
Specifications
Specifications
| Chemical Name or Material | Nicotinuric acid |
| CAS | 583-08-4 |
| Color | White to Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C8H8N2O3 |
| Quantity | 100 mg |
| Solubility Information | DMSO : 100 mg/mL (555.06 mM; Need ultrasonic) ∣H2O : 1.56 mg/mL (8.66 mM; Need ultrasonic) |
| SMILES | O=C(O)CNC(C1=CC=CN=C1)=O |
| Molecular Weight (g/mol) | 180.16 |
| Formula Weight | 180.16 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction