missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nicotinuric Acid, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
Nicotinuric Acid
Specifications
Specifications
| Chemical Name or Material | Nicotinuric Acid |
| CAS | 583-08-4 |
| Analyte or Component Names | Nicotinuric Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C8 H8 N2 O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Glycine, N-(3-pyridinylcarbonyl)-, Glycine, N-nicotinoyl- (6 CI,7 CI,8 CI), (Nicotinoylamino)acetic acid, 2-(Nicotinylamino)acetic acid, NSC 71273, Nicotinoylglycine, Nicotinuric acid, Nicotinylglycine |
| InChI Formula | InChI=1 S/C8H8N2O3/c11-7(12)5-10-8(13)6-2-1-3-9-4-6/h1-4 H,5H2,(H,10,13)(H,11,12) |
| SMILES | OC(=O)CNC(=O)c1cccnc1 |
| IUPAC Name | 2-(pyridine-3-carbonylamino)acetic acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction