missing translation for 'onlineSavingsMsg'
Learn More

Nicotinuric acid, 98+%, Thermo Scientific Chemicals

Product Code. 15424629
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15424629

Brand: Thermo Scientific Alfa Aesar J62215.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Measurement of NUA levels in urine is used to indicate possible elevated nicotinamide adenine dinucleotide (NAD) catabolism. NUA may be used as a reference (standard curve) in analysis of NAD and NUA catabolism. It is an antiproliferative.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Measurement of NUA levels in urine is used to indicate possible elevated nicotinamide adenine dinucleotide (NAD) catabolism. NUA may be used as a reference (standard curve) in analysis of NAD and NUA catabolism. It is an antiproliferative.

Solubility
Soluble in dimethyl sulfoxide, methanol, water, and 1M NH4OH (50 mg/mL).

Notes
Store at -20°C. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 583-08-4
Molecular Formula C8H8N2O3
Molecular Weight (g/mol) 180.163
MDL Number MFCD00023578
InChI Key ZBSGKPYXQINNGF-UHFFFAOYSA-N
Synonym nicotinuric acid, nicotinylglycine, nicotinoylglycine, 2-nicotinamido acetic acid, n-nicotinoylglycine, n-pyridin-3-ylcarbonyl glycine, nicotinoyl-glycine, n-nicotinylglycine, nicotinurate, n-nicotinoyl-glycine
PubChem CID 68499
ChEBI CHEBI:7563
IUPAC Name 2-(pyridine-3-carbonylamino)acetic acid
SMILES C1=CC(=CN=C1)C(=O)NCC(=O)O

Specifications

Color White
Quantity 5 g
Solubility Information Soluble in dimethyl sulfoxide,methanol,water,and 1M NH4OH (50mg/mL).
Formula Weight 180.16
Percent Purity ≥98%
Physical Form Crystalline Powder
Chemical Name or Material Nicotinuric acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.