missing translation for 'onlineSavingsMsg'
Learn More

Nicotinuric acid, 98+%, Thermo Scientific Chemicals

Catalog Number p-7058217
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
583-08-4
C8H8N2O3
180.163
MFCD00023578
ZBSGKPYXQINNGF-UHFFFAOYSA-N
nicotinuric acid, nicotinylglycine, nicotinoylglycine, 2-nicotinamido acetic acid, n-nicotinoylglycine, n-pyridin-3-ylcarbonyl glycine, nicotinoyl-glycine, n-nicotinylglycine, nicotinurate, n-nicotinoyl-glycine
68499
CHEBI:7563
2-(pyridine-3-carbonylamino)acetic acid
C1=CC(=CN=C1)C(=O)NCC(=O)O
This item is not returnable. View return policy
583-08-4
C8H8N2O3
180.163
MFCD00023578
ZBSGKPYXQINNGF-UHFFFAOYSA-N
nicotinuric acid, nicotinylglycine, nicotinoylglycine, 2-nicotinamido acetic acid, n-nicotinoylglycine, n-pyridin-3-ylcarbonyl glycine, nicotinoyl-glycine, n-nicotinylglycine, nicotinurate, n-nicotinoyl-glycine
68499
CHEBI:7563
2-(pyridine-3-carbonylamino)acetic acid
C1=CC(=CN=C1)C(=O)NCC(=O)O

Measurement of NUA levels in urine is used to indicate possible elevated nicotinamide adenine dinucleotide (NAD) catabolism. NUA may be used as a reference (standard curve) in analysis of NAD and NUA catabolism. It is an antiproliferative.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Measurement of NUA levels in urine is used to indicate possible elevated nicotinamide adenine dinucleotide (NAD) catabolism. NUA may be used as a reference (standard curve) in analysis of NAD and NUA catabolism. It is an antiproliferative.

Solubility
Soluble in dimethyl sulfoxide, methanol, water, and 1M NH4OH (50 mg/mL).

Notes
Store at -20°C. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Color White
Quantity 5 g
Solubility Information Soluble in dimethyl sulfoxide,methanol,water,and 1M NH4OH (50mg/mL).
Formula Weight 180.16
Percent Purity ≥98%
Physical Form Crystalline Powder
Chemical Name or Material Nicotinuric acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'