missing translation for 'onlineSavingsMsg'
Learn More

Nicotinoyl chloride hydrochloride, 96%

Product Code. 11448206
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11448206 250 g 250g
11428206 10 g 10g
11438206 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11448206 Supplier Thermo Scientific Alfa Aesar Supplier No. A15317.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 20260-53-1 | C6H5Cl2NO | 178.012 g/mol

Nicotinoyl chloride hydrochloride was used in the preparation of 1-[5-O-tert-butyldimethylsilyl-2-deoxy-2-fluoro-3-O-(3-pyridylcarbonyl)-β-D-ribofuranosyl]uracil, bis(ethylenedithio)tetrathiafulvalene derivatives and 7,16-dinicotinoylated or 7,16-diisonicotinoylated compounds via reaction with tetraaza[14]annulene and its complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nicotinoyl chloride hydrochloride was used in the preparation of 1-[5-O-tert-butyldimethylsilyl-2-deoxy-2-fluoro-3-O-(3-pyridylcarbonyl)-β-D-ribofuranosyl]uracil, bis(ethylenedithio)tetrathiafulvalene derivatives and 7,16-dinicotinoylated or 7,16-diisonicotinoylated compounds via reaction with tetraaza[14]annulene and its complexes.

Solubility
It decomposes in water.

Notes
Moisture sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20260-53-1
Molecular Formula C6H5Cl2NO
Molecular Weight (g/mol) 178.012
MDL Number MFCD00012820
InChI Key MSYBLBLAMDYKKZ-UHFFFAOYSA-N
Synonym nicotinoyl chloride hydrochloride, pyridine-3-carbonyl chloride hydrochloride, nicotinoyl chloride, hydrochloride, 3-pyridinecarbonyl chloride, hydrochloride, 3-pyridinecarbonyl chloride hydrochloride, nicotinic acid chloride hydrochloride, nicotinoylchloride hydrochloride, 3-pyridinecarbonyl chloride, hydrochloride 1:1, nicotinyl chloride hydrochloride, nicotinoyl chloride hcl
PubChem CID 88438
IUPAC Name pyridine-3-carbonyl chloride;hydrochloride
SMILES C1=CC(=CN=C1)C(=O)Cl.Cl

Specifications

Melting Point 149°C to 155°C
Quantity 250 g
UN Number UN3261
Beilstein 3655772
Sensitivity Moisture sensitive
Solubility Information It decomposes in water.
Formula Weight 178.02
Percent Purity 98%
Chemical Name or Material Nicotinoyl chloride hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.