missing translation for 'onlineSavingsMsg'
Learn More

Nicotinic Acid, TRC

Product Code. 30135118
Change view
Click to view available options
Quantity:
50 mg
1 g
10 g
Unit Size:
10g
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135118 10 g 10g
17941165 1 g 1g
17992485 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135118 Supplier Toronto Research Chemicals Supplier No. TRCN42925010G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59-67-6
Molecular Formula C6 H5 N O2
Molecular Weight (g/mol) 123.11
Synonym Nicotinic Acid, Methyl Nicotinate Imp. A (EP), 3-Pyridinecarboxylic acid, Nicotinic acid, 3-Carboxylpyridine, 3-Carboxypyridine, 3-Pyridylcarboxylic acid, Akotin, Apelagrin, CaRo 03, Daskil, E 375, Efacin, Enduracin, Linic, Microvit B3 Promix Niacin, NSC 169454, Niac, Niacin, Niacor, Niaspan, Nicacid, Nicangin, Nico-Span, Nicobid, Nicodelmine, Nicolar, Niconacid, Nicosan 3, Nicotinipca, Nicyl, Nyclin, Pellagrin, Pelonin, SR 4390, Slo-niacin, Vitamin B3, Vitamin B5, Wampocap, β-Pyridinecarboxylic acid
IUPAC Name pyridine-3-carboxylic acid
SMILES OC(=O)c1cccnc1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C6H5NO2/c8-6(9)5-2-1-3-7-4-5/h1-4H,(H,8,9)
Formula Weight 123.032
Chemical Name or Material Niacin (Nicotinic Acid)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.