missing translation for 'onlineSavingsMsg'
Learn More

Nicotinic Acid Riboside, TRC

Product Code. 30008055
Change view
Click to view available options
Quantity:
1 mg
5 mg
25 mg
Unit Size:
1mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008055 25 mg 25mg
30007554 5 mg 5mg
30004503 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008055 Supplier Toronto Research Chemicals Supplier No. TRCN42940025MG
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17720-18-2
Molecular Formula C11 H13 N O6
Molecular Weight (g/mol) 255.22
Synonym Nicotinic riboside, Nicotinic acid ribonucleoside
IUPAC Name 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl]pyridin-1-ium-3-carboxylate
SMILES OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)[n+]2cccc(c2)C(=O)[O-]

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C11H13NO6/c13-5-7-8(14)9(15)10(18-7)12-3-1-2-6(4-12)11(16)17/h1-4,7-10,13-15H,5H2/t7-,8-,9-,10-/m1/s1
Formula Weight 255.0743
Chemical Name or Material beta-Nicotinic acid riboside
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.