missing translation for 'onlineSavingsMsg'
Learn More

Nicotinic acid N-oxide, 99%

Product Code. 10209183 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10209183 500 g Plastic bottle 500g
10435852 5 g Glass bottle 5g
10427730 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10209183 Supplier Thermo Scientific Acros Supplier No. 147785000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2398-81-4 | C6H6NO3 | 140.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2398-81-4
Molecular Formula C6H6NO3
Molecular Weight (g/mol) 140.12
MDL Number MFCD00006201
InChI Key GDILJONVBZVAFH-UHFFFAOYNA-N
Synonym nicotinic acid n-oxide, oxiniacic acid, nicotinic acid 1-oxide, 3-pyridinecarboxylic acid, 1-oxide, nicotinic acid oxide, n-hydroxynicotinic acid, 3-carboxypyridin-1-ium-1-olate, 3-carboxypyridine n-oxide, 3-carboxypyridine 1-oxide, 3-pyridinecarboxylic acid oxide
PubChem CID 16976
IUPAC Name 1-oxidopyridin-1-ium-3-carboxylic acid
SMILES OC(=O)C1=C[NH++]([O-])[CH-]C=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 260.0°C to 262.0°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Quantity 500 g
Merck Index 15, 7040
Formula Weight 139.11
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material Nicotinic acid N-oxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.