missing translation for 'onlineSavingsMsg'
Learn More

Nicotinic Acid, Mikromol™

Product Code. 17953006
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17953006 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17953006 Supplier Mikromol Supplier No. MM0553.000250
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Nicotinic Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Niacin (Nicotinic Acid)
CAS 59-67-6
Analyte or Component Names Niacin (Nicotinic Acid)
Shipping Condition Room Temperature
Molecular Formula C6 H5 N O2
Certifications/Compliance ISO 17034
Synonym Nicotinic Acid, Methyl Nicotinate Imp. A (EP), 3-Pyridinecarboxylic acid, Nicotinic acid, 3-Carboxylpyridine, 3-Carboxypyridine, 3-Pyridylcarboxylic acid, Akotin, Apelagrin, CaRo 03, Daskil, E 375, Efacin, Enduracin, Linic, Microvit B3 Promix Niacin, NSC 169454, Niac, Niacin, Niacor, Niaspan, Nicacid, Nicangin, Nico-Span, Nicobid, Nicodelmine, Nicolar, Niconacid, Nicosan 3, Nicotinipca, Nicyl, Nyclin, Pellagrin, Pelonin, SR 4390, Slo-niacin, Vitamin B3, Vitamin B5, Wampocap, beta-Pyridinecarboxylic acid, Methyl Nicotinate Impurity A
InChI Formula InChI=1S/C6H5NO2/c8-6(9)5-2-1-3-7-4-5/h1-4H,(H,8,9)
SMILES OC(=O)c1cccnc1
IUPAC Name pyridine-3-carboxylic acid
Molecular Weight (g/mol) 123.11
Quantity 250 mg
Formula Weight 123.032 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.