missing translation for 'onlineSavingsMsg'
Learn More

Nicotinic acid, 99%

Product Code. 11470573
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11470573 250 g 250g
11460573 1000 g 1000g
11480573 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11470573 Supplier Thermo Scientific Alfa Aesar Supplier No. A12683.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 59-67-6 | C6H5NO2 | 123.11 g/mol

Vitamin of the B complex with hypolipidemic properties occurring in various animal and plant tissues. Required by the body for the formation of coenzymes NAD and NADP.Nicotinic acid is used as an antidyslipidemic agent. It is also used to improve all lipoproteins well above the vitamin requirement and lowers the total cholesterol, bad LDL-cholesterol, and triglyceride levels. Further, it is used as a food additive in food industries, cosmetics and pharmaceuticals. In addition to this, it is used in medication, nutrient, enriched flours and in electroplating baths.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water. Slightly soluble in ethanol and ethyl ether. Insoluble in most lipid solvents.

Notes
Light sensitive. Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59-67-6
Molecular Formula C6H5NO2
Molecular Weight (g/mol) 123.11
MDL Number MFCD00006391
InChI Key PVNIIMVLHYAWGP-UHFFFAOYSA-N
Synonym nicotinic acid, niacin, 3-pyridinecarboxylic acid, 3-carboxypyridine, wampocap, acidum nicotinicum, apelagrin, pellagrin, akotin, daskil
PubChem CID 938
ChEBI CHEBI:15940
IUPAC Name pyridine-3-carboxylic acid
SMILES OC(=O)C1=CC=CN=C1

Specifications

Melting Point 236°C to 239°C
Density 1.4
Flash Point 293°C (559°F)
Assay Percent Range 99%
Quantity 250 g
Beilstein 109591
Merck Index 14,6525
Solubility Information Soluble in water. Slightly soluble in ethanol and ethyl ether. Insoluble in most lipid solvents.
Formula Weight 123.11
Percent Purity 99%
Chemical Name or Material Nicotinic Acid
Show More Show Less
compliance-icons
Product Identifier
  • Nicotinic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 3-Pyridinecarboxylicacid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.