missing translation for 'onlineSavingsMsg'
Learn More

Nicotinic Acid-13C6 (100 ug/mL in Methanol), TRC

Product Code. 30392197
Change view
Click to view available options
Quantity:
1x1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30392197 1x1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30392197 Supplier Toronto Research Chemicals Supplier No. TRCKIT15871X1ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nicotinic acid-13C6
CAS 1189954-79-7
Molecular Formula 13C6 H5 N O2
Quantity 1x1 mL
Synonym Nicotinic Acid-13C6
InChI Formula InChI=1S/C6H5NO2/c8-6(9)5-2-1-3-7-4-5/h1-4H,(H,8,9)/i1+1,2+1,3+1,4+1,5+1,6+1
SMILES O[13C](=O)[13c]1[13cH][13cH][13cH]n[13cH]1
IUPAC Name (2,3,4,5,6-^{13}C_{5})pyridine-3-carboxylic acid
Molecular Weight (g/mol) 129.07
Formula Weight 129.0522
Concentration or Composition (by Analyte or Components) 100 μg/mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.