missing translation for 'onlineSavingsMsg'
Learn More

Nicotinic Acetylcholine Receptor epsilon Antibody (1H5), Novus Biologicals™

Product Code. 18391508 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18391508 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18391508 Supplier Novus Biologicals Supplier No. H00001145M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Nicotinic Acetylcholine Receptor epsilon Monoclonal antibody specifically detects Nicotinic Acetylcholine Receptor epsilon in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nicotinic Acetylcholine Receptor epsilon
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 1H5
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_000071
Gene Alias acetylcholine receptor subunit epsilon, AchR epsilon subunit, ACHREFCCMS, cholinergic receptor, nicotinic, epsilon, cholinergic receptor, nicotinic, epsilon polypeptide, CMS1D, CMS1E, CMS2A, SCCMS
Host Species Mouse
Immunogen CHRNE (NP_000071, 21 a.a. ~ 130 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. KNEELRLYHHLFNNYDPGSRPVREPEDTVTISLKVTLTNLISLNEKEETLTTSVWIGIDWQDYRLNYSKDDFGGIETLRVPSELVWLPEIVLENNIDGQFGVAYDANVLV
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cancer, Cell Biology, Immunology, Mesenchymal Stem Cell Markers, Myeloid derived Suppressor Cell, Phospho Specific, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 1145
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG3 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.